Order Entry
Belgium
ContactUsLinkComponent
 
VWR® Oxidase Swabs in a Box
Catalog # 942152ZF
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Oxidase Swabs in a Box
Catalog # 942152ZF
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    50 Tests
  • Description:
    Swabs, Oxidase reagent
  • Shelf life:
    360 days
  • Storage temperature:
    2...14 °C

Specifications

About this item

Swabs imprenated with reagent for the detection and verification of bacterial cytochrome oxidase.

  • 2×25 swabs
Caution: *The product(s) below are outside the scope of the EU Directive 2019/904.