Orders LinkContactUsLinkComponent
Cell strainers, Corning®
Supplier:   Corning
Cell strainers, Corning®
Supplier:  Corning
Certificates
Cell strainers, Corning®
Cell strainers, Corning®
3 Options Available Below
Product Details & Documents
Corning® cell strainers are sterile, easy to use devices for rapidly isolating primary cells to consistently obtain a uniform single-cell suspension from tissues. Fits 50 ml Falcon® conical tubes.
  • Strong nylon mesh for optimal performance in a variety of applications
  • Extended lip on strainer enables aseptic handling with forceps
  • Moulded colour-coded polypropylene frame with tab enables easy handling and identification
  • Sterilised by gamma irradiation, non cytotoxic
  • Conveniently accessible in individual packaging
You can use these versatile devices to prepare samples for flow cytometry, single cell suspensions of blood cells, specimens for primary cell cultures and immunogens as well as primary cells from tissues.
Product Family Options
Product Information
  • Availability
    Price
  • 734-2760
    Cell strainer, 40 µm pore size, blue
    Cell strainer, 40 µm pore size, blue
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 106,00 €
    106,00 €
    1 * 50 ST
       106,00 €
  • 734-2761
    Cell strainer, 70 µm pore size, white
    Cell strainer, 70 µm pore size, white
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 106,00 €
    106,00 €
    1 * 50 ST
       106,00 €
  • 734-2762
    Cell strainer, 100 µm pore size, yellow
    Cell strainer, 100 µm pore size, yellow
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 106,00 €
    106,00 €
    1 * 50 ST
       106,00 €