Contact plates, gamma irridiated, triple-bagged, ICR (isolator, cleanroom)
Supplier: MERCK LIFE SCIENCE
Certificates
About this item
ICR/ICRplus contact plates are triple-bagged and gamma irradiated to ensure the highest level of safety when monitoring surfaces in isolators and critical clean rooms for microorganisms.
- Data matrix barcode on each plate for paperless and secure identification of individual plates
- CLOSED and VENT closure options of ICRplus contact plates for safe transportation and variation of incubation conditions
- Convenient room temperature stability allows storage at site of use
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together