ContactUsLinkComponent
Racks for cryogenic vials, TruCool®
Racks for cryogenic vials, TruCool®
Catalog # BCISBCS-222
Supplier:  BIOCISION
undefined
Racks for cryogenic vials, TruCool®
Catalog # BCISBCS-222
Supplier:  BIOCISION
Specifications
  • No. of Holes:
    50 (5×10)
  • Colour:
    Multicolour
  • Description:
    TruCool® cryogenic vial racks
  • W×D×H:
    197×102×220 mm
  • Pk:
    1 SET
Specifications
About this item
These polypropylene cryogenic racks feature a locking mechanism that allow one handed opening for self-standing cryogenic vials.
  • Accommodates both round bottom and self-standing vial formats
  • Racks have A to J row and 1 to 5 column indexing for easy organization
  • Stackable when not in use
Multicolour pack contains one each of blue, green, orange, purple and pink racks.