Order Entry
Belgium
Orders LinkContactUsLinkComponent
Combination pH electrodes, Orion™ Basic

Combination pH electrodes, Orion™ Basic

Supplier:   Thermo Orion
Combination pH electrodes, Orion™ Basic
Supplier:  Thermo Orion
Certificates
Combination pH electrodes, Orion™ Basic
Combination pH electrodes, Orion™ Basic

2 Options Available Below

Product Details & Documents

These pH electrodes are ideal for performing basic pH measurements quickly and reliably. Designed for use with Orion™ Russell meters.

  • Can be used with any meter with BNC input
  • For general purpose applications

Storage requirements: store in Orion pH electrode storage solution 910001.

Six months from date of purchase by buyer or 12 months from date of shipment from seller.

Product Family Options

Product Information

  • Type
    pH
    Temp.
    Material
    Electrolyte
    Ø×L
    Connection
    For
    Availability
    Price
  • ORIO9142BN
    Refillable combination pH electrode with glass body
    Refillable combination pH electrode with glass body
    Orion™ Basic 9142BN
    0 - 14
    0...+80 °C
    Glass
    Refillable
    12×120 mm
    BNC
    Orion™ Russell meters
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 144,00 €
    144,00 €
    1 * 1 ST
       144,00 €

  • ORIO9146BN
    Low maintenance gel combination with epoxy body and fluted guard
    Low maintenance gel combination with epoxy body and fluted guard
    Orion™ Basic 9146BN, with removable bulb guard
    0 - 14
    0...+80 °C
    Epoxy
    Gel
    12×120 mm
    BNC
    Orion™ Russell meters
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €

Accessories

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • pH electrode storage solution, 475 ml bottle
    pH electrode storage solution, 475 ml bottle
    662-0768
    475 mL
    Log in for availability
    1 * 475 mL - 85,40 €

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • Standard, speciality, PerpHecT Ag/AgCl, single junction refillable green and micro pH electrodes, ORP electrodes, 60 ml
    Standard, speciality, PerpHecT Ag/AgCl, single junction refillable green and micro pH electrodes, ORP electrodes, 60 ml
    662-0753
    5
    Log in for availability
    1 * 5 ST - 149,00 €