Orders LinkContactUsLinkComponent
Capillary columns, DB-FFAP
Capillary columns, DB-FFAP
Supplier:   Agilent
Capillary columns, DB-FFAP
Supplier:  Agilent
Certificates
Capillary columns, DB-FFAP
Capillary columns, DB-FFAP
37 Options Available Below
Product Details & Documents
These columns are designed for the analysis of volatile fatty acids and phenols.
  • Highly polar
  • Bonded and cross linked
  • Solvent rinsable
Nitroterephthalic acid modified polyethylene glycol.
Max. temperature: 250 °C
Suitable replacement for OV-351.
Product Family Options
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • HEWL127-3212
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    0,10 µm
    0,10 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 542,00 €
    542,00 €
    1 * 1 ST
       542,00 €
  • HEWL127-3212LTM
    on 7890/6890 LTM module
    on 7890/6890 LTM module
    Capillary
    DB-FFAP
    0,10 µm
    0,10 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HEWL127-32H2
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    0,10 µm
    0,10 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 460,00 €
    460,00 €
    1 * 1 ST
       460,00 €
  • HEWL127-32H2LTM
    on 7890/6890 LTM module
    on 7890/6890 LTM module
    Capillary
    DB-FFAP
    0,10 µm
    0,10 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HEWL122-3212
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    0,25 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 469,00 €
    469,00 €
    1 * 1 ST
       469,00 €
  • HEWL122-3232
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    0,25 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 786,00 €
    786,00 €
    1 * 1 ST
       786,00 €
  • HEWL122-3232E
    on 5" cage
    on 5" cage
    Capillary
    DB-FFAP
    0,25 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 882,00 €
    882,00 €
    1 * 1 ST
       882,00 €
  • HEWL122-3232LTM
    on 7890/6890 LTM module
    on 7890/6890 LTM module
    Capillary
    DB-FFAP
    0,25 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 020,00 €
    2 020,00 €
    1 * 1 ST
       2 020,00 €
  • HEWL122-3233
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    0,50 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 786,00 €
    786,00 €
    1 * 1 ST
       786,00 €
  • HEWL122-3262E
    on 5" cage
    on 5" cage
    Capillary
    DB-FFAP
    0,25 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 550,00 €
    1 550,00 €
    1 * 1 ST
       1 550,00 €
  • HEWL122-3262
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    0,25 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 380,00 €
    1 380,00 €
    1 * 1 ST
       1 380,00 €
  • HEWL122-3263
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    0,50 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 380,00 €
    1 380,00 €
    1 * 1 ST
       1 380,00 €
  • HEWL123-3212
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    0,25 µm
    0,32 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 508,00 €
    508,00 €
    1 * 1 ST
       508,00 €
  • HEWL123-3223
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    0,50 µm
    0,32 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 753,00 €
    753,00 €
    1 * 1 ST
       753,00 €
  • HEWL123-3232
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    0,25 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 850,00 €
    850,00 €
    1 * 1 ST
       850,00 €
  • HEWL123-3232E
    on 5" cage
    on 5" cage
    Capillary
    DB-FFAP
    0,25 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 953,00 €
    953,00 €
    1 * 1 ST
       953,00 €
  • HEWL123-3232LTM
    on 7890/6890 LTM module
    on 7890/6890 LTM module
    Capillary
    DB-FFAP
    0,25 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 100,00 €
    2 100,00 €
    1 * 1 ST
       2 100,00 €
  • HEWL123-3233
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    0,50 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 850,00 €
    850,00 €
    1 * 1 ST
       850,00 €
  • HEWL123-3233E
    on 5" cage
    on 5" cage
    Capillary
    DB-FFAP
    0,50 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 953,00 €
    953,00 €
    1 * 1 ST
       953,00 €
  • HEWL123-3233LTM
    on 7890/6890 LTM module
    on 7890/6890 LTM module
    Capillary
    DB-FFAP
    0,50 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 100,00 €
    2 100,00 €
    1 * 1 ST
       2 100,00 €
  • HEWL123-3234
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    1,00 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 850,00 €
    850,00 €
    1 * 1 ST
       850,00 €
  • HEWL123-3234LTM
    on 7890/6890 LTM module
    on 7890/6890 LTM module
    Capillary
    DB-FFAP
    1,00 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 100,00 €
    2 100,00 €
    1 * 1 ST
       2 100,00 €
  • HEWL123-3253
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    0,50 µm
    0,32 mm
    50 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 350,00 €
    1 350,00 €
    1 * 1 ST
       1 350,00 €
  • HEWL123-3262
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    0,25 µm
    0,32 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 510,00 €
    1 510,00 €
    1 * 1 ST
       1 510,00 €
  • HEWL123-3263
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    0,50 µm
    0,32 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 510,00 €
    1 510,00 €
    1 * 1 ST
       1 510,00 €
  • HEWL123-3264
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    1,00 µm
    0,32 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 510,00 €
    1 510,00 €
    1 * 1 ST
       1 510,00 €
  • HEWL124-3232
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    0,85 µm
    0,45 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 943,00 €
    943,00 €
    1 * 1 ST
       943,00 €
  • HEWL125-32H2
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    1,00 µm
    0,53 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 431,00 €
    431,00 €
    1 * 1 ST
       431,00 €
  • HEWL125-3212
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    1,00 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 547,00 €
    547,00 €
    1 * 1 ST
       547,00 €
  • HEWL125-3217
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    0,50 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 547,00 €
    547,00 €
    1 * 1 ST
       547,00 €
  • HEWL125-3217LTM
    on 7890/6890 LTM module
    on 7890/6890 LTM module
    Capillary
    DB-FFAP
    0,50 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HEWL125-3231
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    0,25 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 943,00 €
    943,00 €
    1 * 1 ST
       943,00 €
  • HEWL125-3232
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    1,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 943,00 €
    943,00 €
    1 * 1 ST
       943,00 €
  • HEWL125-3232E
    on 5" cage
    on 5" cage
    Capillary
    DB-FFAP
    1,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 060,00 €
    1 060,00 €
    1 * 1 ST
       1 060,00 €
  • HEWL125-3233
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    1,50 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 943,00 €
    943,00 €
    1 * 1 ST
       943,00 €
  • HEWL125-3237
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    0,50 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 943,00 €
    943,00 €
    1 * 1 ST
       943,00 €
  • HEWL125-3262
    on 7" cage
    on 7" cage
    Capillary
    DB-FFAP
    1,00 µm
    0,53 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 590,00 €
    1 590,00 €
    1 * 1 ST
       1 590,00 €