pH/mV/°C meter, bench, HQd (HQ411D)
: Hach
This advanced laboratory meter is ideal for testing critical water quality parameters. The meter connects with smart probes (IntelliCAL™) that automatically recognise the testing parameter, calibration history, and method settings to minimise errors and setup time. The instrument is designed to withstand years of continuous lab use.
- Single input channel for pH, ORP, mV, and temperature: For use with any IntelliCAL™ smart pH or ORP electrode
- Simple user interface that does not require manuals or training to operate
- Guided calibration and check standard routines reduce calibration errors
- Internal USB ports simplify data transfer, peripherals, and software updates and connection to PCs printer and keyboards
- Fully GLP compliant communication and documentation
- Ultra-bright screen and large font size for easy to read results
: Supplied with four batteries, universal power adapter and with or without electrode holder. IntelliCAL™ electrodes need to be ordered separately.
- Catalog No:
- 665-0380
- 662-1981
- Model:
- HQ411D
- HQ411D
- pH range:
- 0 - 14
- 0 - 14
- pH resolution:
- 0,1/ 0,01/ 0,001
- 0,1/ 0,01/ 0,001
- pH accuracy:
- ±0,002
- ±0,002
- Automatic buffer recognition:
- Colour-coded: 4.01, 7.00, 10.01 pH;
IUPAC: 1.679, 4.005, 7.000, 10.012, 12.45 pH
DIN: 1.09, 4.65, 9.23
User-defined custom buffer sets - Colour-coded: 4.01, 7.00, 10.01 pH;
IUPAC: 1.679, 4.005, 7.000, 10.012, 12.45 pH
DIN: 1.09, 4.65, 9323
User-defined custom buffer sets
- mV range:
- ±1500
- ±1500
- mV resolution:
- 0,1/ 0,01/ 0,001
- 0,1/ 0,01/ 0,001
- mV accuracy:
- ±0,1
- ±0,1
- Temperature range:
- −10...+110 °C
- −10...+110 °C
- Temperature resolution:
- 0,1 °C
- 0,1 °C
- Temperature accuracy:
- ± 0,3 °C
- ± 0,3 °C
- Temperature compensation:
- Automatic, off, manual
- Automatic, off, manual
- Display:
- Backlit graphical LCD
- Backlit graphical LCD
- Power supply:
- 230 V with power supply or 4 × AA Alkaline batteries
- 230 V with power supply or 4 × AA Alkaline batteries
- Data logging capacity:
- 500 results
- 500 results
- Connections:
- 1 × M12 digital for IntelliCAL™ electrodes
- 1 × M12 digital for IntelliCAL™ electrodes
- Outputs:
- USB
- USB
- W×D×H:
- 175×234×86 mm
- 175×234×86 mm
- Weight:
- 0,85 kg
- 0,85 kg
Electrodes
Frequently Bought Together