Orders LinkContactUsLinkComponent
Dip slides
Dip slides
Dip slides
Supplier:  LIOFILCHEM
Certificates
Dip slides
13 Options Available Below
Product Details & Documents
Flexible contact slide for microbiology screening of surfaces.
  • ISO 18593
Documents
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • LIOF525322
    Contact slide 9 PCA + TTC + Neutralising×2
    Contact slide 9 PCA + TTC + Neutralising×2
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 51,90 €
    51,90 €
    1 * 20 ST
       51,90 €
  • LIOF525292
    Contact slide Chrom 1 Salmonella
    Contact slide Chrom 1 Salmonella
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 59,40 €
    59,40 €
    1 * 20 ST
       59,40 €
  • LIOF525392
    Contact slide Chrom 2
    Contact slide Chrom 2
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 59,40 €
    59,40 €
    1 * 20 ST
       59,40 €
  • LIOF525462
    Contact slide Chrom 3
    Contact slide Chrom 3
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 59,40 €
    59,40 €
    1 * 20 ST
       59,40 €
  • LIOF50021
    Dipslide Dermatest
    Dipslide Dermatest
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 26,00 €
    26,00 €
    1 * 10 ST
       26,00 €
  • LIOF53531
    Enterobact/Strepto + Neutr
    Enterobact/Strepto + Neutr
    120
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 281,00 €
    281,00 €
    1 * 120 ST
       281,00 €
  • LIOF53528
    Pseudomonas/ Lev.Moisis + N
    Pseudomonas/ Lev.Moisis + N
    120
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 281,00 €
    281,00 €
    1 * 120 ST
       281,00 €
  • LIOF525262
    Total / Enterobacteria with neutraliser
    Total / Enterobacteria with neutraliser
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 51,90 €
    51,90 €
    1 * 20 ST
       51,90 €
  • LIOF53530
    Total with Neutr/ Staph
    Total with Neutr/ Staph
    120
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 281,00 €
    281,00 €
    1 * 120 ST
       281,00 €
  • LIOF53562
    Total/Coliform + Neutr
    Total/Coliform + Neutr
    120
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 281,00 €
    281,00 €
    1 * 120 ST
       281,00 €
  • LIOF53526
    Total/Enterobac + Neutraliser
    Total/Enterobac + Neutraliser
    120
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 281,00 €
    281,00 €
    1 * 120 ST
       281,00 €
  • LIOF53527
    Total/Lev Moisis + Ne
    Total/Lev Moisis + Ne
    120
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 281,00 €
    281,00 €
    1 * 120 ST
       281,00 €
  • LIOF500232
    Uritest N
    Uritest N
    20 Tests
    Log in for availability
    Log in for availability
    1 * 20 Tests
    1 * 20 Tests - 28,50 €
    28,50 €
    1 * 20 Tests
       28,50 €