ContactUsLinkComponent
Calibration weights, OIML R111, class M2
Calibration weights, OIML R111, class M2
Catalog # ZWIEE201.12.11
Supplier:  ZWIEBEL
Calibration weights, OIML R111, class M2
Catalog # ZWIEE201.12.11
Supplier:  ZWIEBEL
Specifications
  • Weight:
    1 kg
  • Pk:
    1
  • Tolerance:
    160 ± mg
  • Type:
    Class M2 weights with calibration certificate
Specifications
About this item
Weights made from brass. M2 weights are designed to calibrate masses of a lower accuracy class and be used with class II and III weighing instruments.
  • Ultra-compact and easy to handle
  • Weights are available as individual units or in sets (wooden box/base)