Orders LinkContactUsLinkComponent
Capillary columns, DB-17
Capillary columns, DB-17
Supplier:   Agilent
Capillary columns, DB-17
Supplier:  Agilent
Certificates
Capillary columns, DB-17
Capillary columns, DB-17
41 Options Available Below
Product Details & Documents
These columns contain (50%-Phenyl)-methylpolysiloxane. They are excellent for confirmation analysis.
  • Solvent rinsable
  • Mid polarity
  • Equivalent to USP phase G3
  • Bonded and cross-linked
Max. temperature: 40 to 280/300 °C
Product Family Options
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • HEWL127-1712
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,10 µm
    0,10 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 517,00 €
    517,00 €
    1 * 1 ST
       517,00 €
  • HEWL127-1712LTM
    on LTM module
    on LTM module
    Capillary
    DB-17
    0,10 µm
    0,10 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HEWL127-1713
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,20 µm
    0,10 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 517,00 €
    517,00 €
    1 * 1 ST
       517,00 €
  • HEWL127-1722
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,10 µm
    0,10 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 788,00 €
    788,00 €
    1 * 1 ST
       788,00 €
  • HEWL121-1722
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,18 µm
    0,18 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 647,00 €
    647,00 €
    1 * 1 ST
       647,00 €
  • HEWL121-1722LTM
    on LTM module
    on LTM module
    Capillary
    DB-17
    0,18 µm
    0,18 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 900,00 €
    1 900,00 €
    1 * 1 ST
       1 900,00 €
  • HEWL121-1723
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,30 µm
    0,18 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 647,00 €
    647,00 €
    1 * 1 ST
       647,00 €
  • HEWL122-1712
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,25 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 459,00 €
    459,00 €
    1 * 1 ST
       459,00 €
  • HEWL122-1713
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,50 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 459,00 €
    459,00 €
    1 * 1 ST
       459,00 €
  • HEWL122-1713E
    on 5" cage
    on 5" cage
    Capillary
    DB-17
    0,50 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 515,00 €
    515,00 €
    1 * 1 ST
       515,00 €
  • HEWL122-1731
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,15 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 771,00 €
    771,00 €
    1 * 1 ST
       771,00 €
  • HEWL122-1731E
    on 5" cage
    on 5" cage
    Capillary
    DB-17
    0,15 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 865,00 €
    865,00 €
    1 * 1 ST
       865,00 €
  • HEWL122-1732
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,25 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 771,00 €
    771,00 €
    1 * 1 ST
       771,00 €
  • HEWL122-1732E
    on 5" cage
    on 5" cage
    Capillary
    DB-17
    0,25 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 865,00 €
    865,00 €
    1 * 1 ST
       865,00 €
  • HEWL122-1732LTM
    on LTM module
    on LTM module
    Capillary
    DB-17
    0,25 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 000,00 €
    2 000,00 €
    1 * 1 ST
       2 000,00 €
  • HEWL122-1733
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,50 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 771,00 €
    771,00 €
    1 * 1 ST
       771,00 €
  • HEWL122-1762
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,25 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 360,00 €
    1 360,00 €
    1 * 1 ST
       1 360,00 €
  • HEWL123-1711
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,15 µm
    0,32 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 498,00 €
    498,00 €
    1 * 1 ST
       498,00 €
  • HEWL123-1712
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,25 µm
    0,32 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 498,00 €
    498,00 €
    1 * 1 ST
       498,00 €
  • HEWL123-1713
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,50 µm
    0,32 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 498,00 €
    498,00 €
    1 * 1 ST
       498,00 €
  • HEWL123-1731
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,15 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 833,00 €
    833,00 €
    1 * 1 ST
       833,00 €
  • HEWL123-1732
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,25 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 833,00 €
    833,00 €
    1 * 1 ST
       833,00 €
  • HEWL123-1732E
    on 5" cage
    on 5" cage
    Capillary
    DB-17
    0,25 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 934,00 €
    934,00 €
    1 * 1 ST
       934,00 €
  • HEWL123-1732LTM
    on LTM module
    on LTM module
    Capillary
    DB-17
    0,25 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 080,00 €
    2 080,00 €
    1 * 1 ST
       2 080,00 €
  • HEWL123-1733
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,50 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 833,00 €
    833,00 €
    1 * 1 ST
       833,00 €
  • HEWL123-1733E
    on 5" cage
    on 5" cage
    Capillary
    DB-17
    0,50 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 934,00 €
    934,00 €
    1 * 1 ST
       934,00 €
  • HEWL123-1762
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,25 µm
    0,32 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 480,00 €
    1 480,00 €
    1 * 1 ST
       1 480,00 €
  • HEWL125-1704
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    2,00 µm
    0,53 mm
    5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 326,00 €
    326,00 €
    1 * 1 ST
       326,00 €
  • HEWL125-1711
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,25 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 509,00 €
    509,00 €
    1 * 1 ST
       509,00 €
  • HEWL125-1712
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    1,00 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 509,00 €
    509,00 €
    1 * 1 ST
       509,00 €
  • HEWL125-1712LTM
    on LTM module
    on LTM module
    Capillary
    DB-17
    1,00 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 730,00 €
    1 730,00 €
    1 * 1 ST
       1 730,00 €
  • HEWL125-1713
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    1,50 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 509,00 €
    509,00 €
    1 * 1 ST
       509,00 €
  • HEWL125-1713LTM
    on LTM module
    on LTM module
    Capillary
    DB-17
    1,50 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HEWL125-1717
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,50 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 509,00 €
    509,00 €
    1 * 1 ST
       509,00 €
  • HEWL125-1731
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,25 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 924,00 €
    924,00 €
    1 * 1 ST
       924,00 €
  • HEWL125-1732
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    1,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 924,00 €
    924,00 €
    1 * 1 ST
       924,00 €
  • HEWL125-1732E
    on 5" cage
    on 5" cage
    Capillary
    DB-17
    1,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 040,00 €
    1 040,00 €
    1 * 1 ST
       1 040,00 €
  • HEWL125-1732LTM
    on LTM module
    on LTM module
    Capillary
    DB-17
    1,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 190,00 €
    2 190,00 €
    1 * 1 ST
       2 190,00 €
  • HEWL125-1733
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    1,50 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 924,00 €
    924,00 €
    1 * 1 ST
       924,00 €
  • HEWL125-1737
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    0,50 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 924,00 €
    924,00 €
    1 * 1 ST
       924,00 €
  • HEWL125-1762
    on 7" cage
    on 7" cage
    Capillary
    DB-17
    1,00 µm
    0,53 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 490,00 €
    1 490,00 €
    1 * 1 ST
       1 490,00 €