Order Entry
Belgium
ContactUsLinkComponent
Wipes for general applications, Multitex® Extra
Wipes for general applications, Multitex® Extra
Catalog # ZVGA11818-00
undefined
Wipes for general applications, Multitex® Extra
Catalog # ZVGA11818-00

Specifications

  • Packed:
    1 roll of 190 m
  • Colour:
    White
  • Description:
    Polishing and cleaning cloth roll, perforated
  • L×W:
    400×300 mm
  • Pk:
    1 Roll

Specifications

About this item

These high-quality viscose fleece wipes with textile characteristics are suitable for wet and dry cleaning. The cloths are very soft and yet lint-free, they can absorb large amounts of liquid, remain tear-resistant and meet food safety requirements. Multitex® cloths can be washed out after cleaning, wrung out, and reused.

  • Solvent resistant and fibre-free
  • Wipe can take up large amounts of liquid thanks to its coarse hole structure
  • High tear resistance, even when wet

Multitex® cloths can be washed out after cleaning, wrung out, and reused. Ideal for use in sensitive work areas, such as precision mechanics, electrical and automotive industries, and in paint shops.

Colour: White