Combination glove box, fibreglass, Protector®
About this item
This combination glove box with fibreglass lining provides inlet and outlet HEPA or ULPA filtration and a leak-tight physical barrier to protect the operator from exposure to potentially hazardous materials and perform work with contamination-sensitive materials under vacuum or in an inert environment.
- LCD display provides real time monitoring of volume dilution rates in cubic feet per minute (CFM), air changes/minute (ACM), main chamber pressure (inches of water), and transfer chamber in inches of mercury and mBar
- Large viewing area (91×70 cm) provides maximum visibility; viewing window is ³/₈" thick, laminated safety glass
- Space-saving inner and outer transfer chamber doors that pivot upward, are counterbalanced and equipped with quick-latches
- Interior right-hand side wall-mounted electrical receptacle with 10 amp maximum (230 volt models) circuit load
- Switches for light and electrical duplex
- Two 8" int.Ø, epoxy-coated aluminum double-grooved glove ports mounted 17" apart, with neoprene gaskets
- Two 25-watt fluorescent lights.
- Dry-powder epoxy-coated, 18 and 20 gauge steel exterior panels
The glove boxe features two internal valves to convert the glove box from a flow-through filtered box to an inert atmosphere glove box.
Inlet and outlet HEPA filters are 99,99% efficient on 0,3 micron particulates and are rated for 70 CFM at <0,90" static pressure.
Delivery information: Supplied with a pair of neoprene gloves (size 9³/₄, 762 mm length), 208 to 230 V outlet and a cable with EU or UK-plug. For additional accessories please contact your local Avantor customer service centre.
2 Options Available Below
Product Details & Documents
This combination glove box with fibreglass lining provides inlet and outlet HEPA or ULPA filtration and a leak-tight physical barrier to protect the operator from exposure to potentially hazardous materials and perform work with contamination-sensitive materials under vacuum or in an inert environment.
- LCD display provides real time monitoring of volume dilution rates in cubic feet per minute (CFM), air changes/minute (ACM), main chamber pressure (inches of water), and transfer chamber in inches of mercury and mBar
- Large viewing area (91×70 cm) provides maximum visibility; viewing window is ³/₈" thick, laminated safety glass
- Space-saving inner and outer transfer chamber doors that pivot upward, are counterbalanced and equipped with quick-latches
- Interior right-hand side wall-mounted electrical receptacle with 10 amp maximum (230 volt models) circuit load
- Switches for light and electrical duplex
- Two 8" int.Ø, epoxy-coated aluminum double-grooved glove ports mounted 17" apart, with neoprene gaskets
- Two 25-watt fluorescent lights.
- Dry-powder epoxy-coated, 18 and 20 gauge steel exterior panels
The glove boxe features two internal valves to convert the glove box from a flow-through filtered box to an inert atmosphere glove box.
Inlet and outlet HEPA filters are 99,99% efficient on 0,3 micron particulates and are rated for 70 CFM at <0,90" static pressure.
Delivery information: Supplied with a pair of neoprene gloves (size 9³/₄, 762 mm length), 208 to 230 V outlet and a cable with EU or UK-plug. For additional accessories please contact your local Avantor customer service centre.
Documents
TDS_Protector CA_Combo Glove Boxes#D643
Product specification
Specifications for Product Family
Specifications
- Catalog No:
- LBCP5080065
- LBCP5080070
- Model:
- Protetor®
- Protetor®
- Observation window (W×H):
- 914,5×701 mm
- 914,5×701 mm
- W×D×H ext.:
- 1623×874×1161 mm
- 1623×874×1161 mm
- Weight:
- 227 kg
- 227 kg
Specifications
Product Family Options
Product Information
- AvailabilityPrice
- LBCP5080065Glove box, Protector®, fibre glass, with 2 glove ports, UK-plugGlove box, Protector®, fibre glass, with 2 glove ports, UK-plugLog in for availability1 * 1 ST54 280,00 €Specifications:
More
Specifications
Cat. No.LBCP5080065ModelProtetor®Observation window (W×H)914,5×701 mmW×D×H ext.1623×874×1161 mmWeight227 kg - LBCP5080070Glove box, Protector®, fibre glass, with 2 glove ports, EU-plugGlove box, Protector®, fibre glass, with 2 glove ports, EU-plugLog in for availability1 * 1 ST54 280,00 €Specifications:
More
Specifications
Cat. No.LBCP5080070ModelProtetor®Observation window (W×H)914,5×701 mmW×D×H ext.1623×874×1161 mmWeight227 kg
Accessories
Product Information
- Catalog #AvailabilityPrice
- Base stand, stationary, with adjustable height (83,8 - 101,6 cm) and leveling feet, 1524×1016×838 mmBase stand, stationary, with adjustable height (83,8 - 101,6 cm) and leveling feet, 1524×1016×838 mm135-1085Log in for availability1 * 1 ST - 3 770,00 €
Product Information
- Catalog #AvailabilityPrice
- Base stand, mobile, with adjustable height (83,8 - 101,6 cm) and PU casters, 1524×1016×838 mmBase stand, mobile, with adjustable height (83,8 - 101,6 cm) and PU casters, 1524×1016×838 mm135-1098Log in for availability1 * 1 ST - 3 770,00 €