Specifications
- Antibody type:Primary
- Antigen name:Complement C5
- Antigen symbol:C5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Sheep
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western blot:Yes
- Cross adsorption:No
- Format:Purified IgG
- Form:Clear, colorless liquid, 0.2 µm filtered
- Antigen synonyms:anaphylatoxin C5|C5
- Storage buffer:10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Storage temperature:2...8 °C
- Concentration:> 4,5 mg/ml (E 1% at 280 nm = 13,0)
- Shipping temperature:2-8 °C
- Immunogen:Human Complement C5
- Tested applications:Fluorescence-Activated Cell Sorting, Immunodiffusion, Labeling
- Purification:Affinity purified using solid phase Human Complement C5
- Size:0,5 mg
- Pk:0,5 mg
Specifications
About this item
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy. Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.
- Origin: US
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Type: Primary
Antigen: C5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human