Order Entry
Belgium
ContactUsLinkComponent
Anti-IgG Donkey Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Anti-IgG Donkey Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # IMMRIR1243
Supplier:  ImmunoReagents
Anti-IgG Donkey Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # IMMRIR1243
Supplier:  ImmunoReagents

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    TRITC (Tetramethylrhodamine Isothiocyanate)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Donkey
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rabbit
  • Western blot:
    Yes
  • Cross adsorption:
    Yes
  • Format:
    Purified IgG
  • Form:
    Lyophilised
  • Storage buffer:
    10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 1% (w/v) BSA, Protease/IgG free
  • Storage temperature:
    2...8 °C
  • Concentration:
    1,0 mg/ml (E 1% at 280 nm = 13,0)
  • Shipping temperature:
    2...8 °C
  • Immunogen:
    Rabbit IgG
  • Tested applications:
    Immunomicroscopy
  • Purification:
    Affinity purified using solid phaseRabbit IgG
  • Size:
    1,0 mg
  • Pk:
    2 mg

Specifications

About this item

TRITC (Tetramethylrhodamine isothiocyanate) is a popular rhodamine derivative pink in Colour but emits a red-orange Colour upon Emission at 580 nm. TRITC is frequently paired with FITC in double labeling experiments and conjugated antibodies are often used in immunofluorescence microscopy, flow cytometry, FLISA (fluorescent ELISA) and high throughput screening assays. These conjugates are best excited with the 532 nm spectral line of the He-Ne laser and are also recommended as a second Colour detection reagent in in situ hybridization applications. (Excitation/Emission = 555 nm/580 nm Emission Colour = Orange to Red (Similar Dyes: Alexa Fluor 555, Cy3, DyLight 550).

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002.
Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Rabbit