Anti-STAT 4 Goat Polyclonal Antibody
USBIS7972-02J1
- Antibody type:Primary
- Antigen symbol:STAT 4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Amino acid number:369 to 383
- Storage buffer:trissaline, 0,02% sodium azide, pH 7,3, 0,5% BSA.
- Sequence:TLSNRRFVLCGTNVK
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: ELISA: 1:1000; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: STAT 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human