Anti-Glutamate Transporter, Aspartate EAAT1 Rabbit polyclonal antibody
USBIG4003-90W
- Antibody type:Primary
- Antigen name:Glutamate Transporter, Aspartate EAAT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Rabbit,Human,Rat,Mouse
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS, 5% BSA, 0,05% thimerosal, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml (upon reconstitution)
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 0.5-1.0 ?g/ml
Type: Primary
Antigen: Glutamate Transporter, Aspartate EAAT1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rabbit, Rat