Order Entry
Belgium
ContactUsLinkComponent
Anti-Complement C3a, C3a des-Arg Mouse Monoclonal Antibody [clone: 6A529]
Anti-Complement C3a, C3a des-Arg Mouse Monoclonal Antibody [clone: 6A529]
Catalog # USBIC7850-13G
Supplier:  US Biological
Anti-Complement C3a, C3a des-Arg Mouse Monoclonal Antibody [clone: 6A529]
Catalog # USBIC7850-13G
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Complement C3a, C3a des-Arg
  • Clonality:
    Monoclonal
  • Clone:
    6A529
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,1% BSA, 0,02% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 1:50

Type: Primary
Antigen: Complement C3a, C3a des-Arg
Clonality: Monoclonal
Clone: 6A529
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human