Anti-CAD Rabbit polyclonal antibody unconjugated
Catalog # USBIC0106-05
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase
- Antigen symbol:CAD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7.0, 0,01% thimerosal, 10% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:500-1:3000; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: CAD (carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human