Anti-ASS1 Rabbit polyclonal antibody unconjugated
Catalog # USBIA3550-66A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Argininosuccinate Synthase 1
- Antigen symbol:ASS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:internal
- Form:liquid
- Antigen synonyms:CTLN1
- Storage buffer:PBS, 0,122% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100
Type: Primary
Antigen: ASS1 (argininosuccinate synthase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse