Anti-AMP Activated Protein Kinase alpha1/2 Mouse Monoclonal Antibody [clone: 9E22]
Catalog # USBIA1475-01U
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:AMP Activated Protein Kinase alpha 1/2
- Antigen symbol:AMPKA1/2
- Clonality:Monoclonal
- Clone:9E22
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Drosophila,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:100-1:1000
Type: Primary
Antigen: AMP Activated Protein Kinase alpha1/2
Clonality: monoclonal
Clone: 9E+22
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Drosophila, Human, Mouse, Rat