Anti-Siglec-H Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 730407]
Catalog # USBI145607
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Sialic Acid Binding Ig-like Lectin H
- Antigen symbol:SIGLECH
- Clonality:Monoclonal
- Clone:730407
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG1
- Reactivity:Mouse
- Form:liquid
- Amino acid number:1 to 269
- Storage buffer:PBS, 0,5% BSA, 0,1% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Do not freeze!
Type: Primary
Antigen: Siglec-H
Clonality: monoclonal
Clone: 730407
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse