Anti-GRIN2A Rabbit Polyclonal Antibody
Catalog # USBI136905
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Glutamate receptor, ionotropic, N-methyl D-aspartate 2A
- Antigen symbol:GRIN2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Format:Serum
- Form:lyophilized powder
- Amino acid number:1253 to 1391
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000-1:2000; Immunochemistry: 1:1000
Type: Primary
Antigen: GRIN2A (glutamate receptor, ionotropic, N-methyl D-aspartate 2A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat