Anti-MCEE Rabbit Polyclonal Antibody
200 µl
Catalog # USBI038268
Supplier: US Biological
Jump to:
Product Details & Documents
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Type: Primary
Antigen: MCEE (methylmalonyl CoA epimerase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Documents
Specifications
- Size:200 µl
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by Protein A chromatography
- Antibody type:Primary
- Epitope:C-terminal
- Form:liquid
- Amino acid number:135 to 164
- Isotype:IgG
- Storage temperature:Cool
- Host:Rabbit
- Shipping temperature:Ice
- Antigen name:methylmalonyl CoA epimerase
- Cat. No.:USBI038268
- Reactivity:Human|Mouse
- Conjugation:Unconjugated
- Antigen symbol:MCEE
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



