Anti-BRIP1 Rabbit Polyclonal Antibody
100 µg
ROCK600-401-458
This affinity purified antibody has been tested for use in ELISA and western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 105 -140 kDa in size corresponding to isoforms of BACH1 Protein by western blotting.
Recommended Dilutions: ELISA: 1:10000 - 1:44000; Western Blot: 1:500- 1:2000;
Type: Primary
Antigen: BRIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Size:100 µg
- Western blot:Yes
- Clonality:Polyclonal
- ImmunoPrecipitation:Yes
- Purification:affinity-purified
- Antibody type:Primary
- Epitope:internal
- Form:Liquid (sterile filtered)
- Amino acid number:92 to 104
- Storage temperature:Cool
- Host:Rabbit
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Cross adsorption:none
- Shipping temperature:Ice
- Antigen name:BRCA1 interacting protein C-terminal helicase 1
- Cat. No.:ROCK600-401-458
- Reactivity:Human
- Format:Antibody fraction
- Gene ID:83990
- Antigen synonyms:BACH1|FANCJ|OF
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a recombinant Protein corresponding to amino acids 92-104 of isoform 1 of human BACH1 Protein.
- Conjugation:Unconjugated
- Antigen symbol:BRIP1
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- ELISA:Yes



