Anti-CDK1 Mouse Polyclonal Antibody (Unconjugated) [clone: POH-1]
Catalog # ROCK200-301-160
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase 1
- Antigen symbol:CDK1
- Clonality:Monoclonal
- Clone:POH-1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:983
- Antigen synonyms:P34CDC2|CDC2|CDC28A
- Storage buffer:0,02 M Potassium Phosphate, 0,5 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:This Protein A purified monoclonal antibody was produced by repeated immunizations with recombinant human p34 cdc2 fusion Protein.
- Purification:Protein A purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Primary Mouse Anti-cdc2 (p34) Reacts with Human, Mouse, Rat
Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:1000; Immunoprecipitation: 1:100;
Type: Primary
Antigen: CDK1
Clonality: Polyclonal
Clone: POH-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse, Rat, Human