Order Entry
Belgium
ContactUsLinkComponent
Anti-c-ErbB2/c-Neu (Ab-6) (OD-3) Mouse Monoclonal Antibody [clone: OD-3]
Anti-c-ErbB2/c-Neu (Ab-6) (OD-3) Mouse Monoclonal Antibody [clone: OD-3]
Catalog # OP67-100
Anti-c-ErbB2/c-Neu (Ab-6) (OD-3) Mouse Monoclonal Antibody [clone: OD-3]
Catalog # OP67-100

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    V-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2
  • Antigen symbol:
    C-ERBB2
  • Clonality:
    Monoclonal
  • Clone:
    OD-3
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: c-ErbB2/c-Neu (Ab-6) (OD-3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human