Anti-Glial Fibrillary Acidic Protein Cocktail (SMI-22) Mouse Monoclonal Antibody [clone: SMI-22]
Catalog # NE1015-100
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen name:Glial Fibrillary Acidic Protein Cocktail
- Clonality:Monoclonal
- Clone:SMI-22
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Glial Fibrillary Acidic Protein Cocktail (SMI-22)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: