Order Entry
Belgium
ContactUsLinkComponent
Anti-COL1 Rabbit Polyclonal Antibody
Anti-COL1 Rabbit Polyclonal Antibody
Catalog # 234167-500
Anti-COL1 Rabbit Polyclonal Antibody
Catalog # 234167-500

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    collagen, type I
  • Antigen symbol:
    COL1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    500 µl
  • Pk:
    500 µl

Specifications

About this item

Type: Primary
Antigen: COL1 (collagen, type I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: