Anti-ENC1 Goat polyclonal antibody unconjugated
Catalog # EVBIEB11784
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:ectodermal-neural cortex 1 (with BTB domain)
- Antigen symbol:ENC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: ENC1 (ectodermal-neural cortex 1 (with BTB domain))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: