Anti-Alpha-synuclein (NACP140) Goat Polyclonal Antibody
Catalog # EVBIEB11714
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:Synuclein alpha
- Antigen symbol:SNCA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Pig,Human
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Antigen synonyms:PARK1|PARK4|NACP|PD1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Alpha-synuclein (NACP140)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Pig