Order Entry
Belgium
ContactUsLinkComponent
Anti-X-arrestin / arrestin 3 Goat Polyclonal Antibody
Anti-X-arrestin / arrestin 3 Goat Polyclonal Antibody
Catalog # EVBIEB07599
Supplier:  Everest Biotech
Anti-X-arrestin / arrestin 3 Goat Polyclonal Antibody
Catalog # EVBIEB07599
Supplier:  Everest Biotech

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    X-arrestin
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Antigen synonyms:
    arrestin 3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: X-arrestin / arrestin 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human