Anti-VRL1 Rabbit Polyclonal Antibody
100 µg
Catalog # BSBTPA1977
Supplier: Boster Bio
Jump to:
Product Details & Documents
Rabbit IgG polyclonal antibody for Transient receptor potential cation channel subfamily V member 2(TRPV2) detection. Tested with WB, IHC-P, IHC-F in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: VRL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Documents
PA1977
Datasheet
Specifications
- Size:100 µg
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by immunogen affinity purification
- Antibody type:Primary
- Epitope:N-Terminal
- Form:Lyophilized
- Amino acid number:42 to 624
- Isotype:IgG
- Storage temperature:–20 °C
- Host:Rabbit
- Sequence:QFQGEDRKFAPQIRVNLNYRK
- Cross adsorption:No
- Shipping temperature:Ice
- Antigen name:Transient Receptor Potential Cation Channel Subfamily V member 2
- Cat. No.:BSBTPA1977
- Reactivity:Human
- Antigen synonyms:VRL1|VRL|VRL-1
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human VRL1 (42-624aa QFQGEDRKFAPQIRVNLNYRK).
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:TRPV2
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



