Order Entry
Belgium
ContactUsLinkComponent
Anti-BST2 Rabbit Polyclonal Antibody
Anti-BST2 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-27349
Supplier:  Novus Biologicals
undefined
Anti-BST2 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-27349
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    bone marrow stromal cell antigen 2
  • Antigen symbol:
    BST2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    684
  • Antigen synonyms:
    CD317|TETHERIN
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: BST2 (bone marrow stromal cell antigen 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human