Anti-SERF1A Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The SERF1A Antibody from Novus Biologicals is a rabbit polyclonal antibody to SERF1A. This antibody reacts with human. The SERF1A Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:400
Type: Primary
Antigen: SERF1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- NOVUNBP1-78393
- NOVUNBP1-78393SS
- Antigen symbol:
- SERF1A
- SERF1A
- Antigen name:
- Small EDRK-rich Factor 1A
- Small EDRK-rich Factor 1A
- Antigen synonyms:
- H4F5|SERF1|FAM2A|4F5|SMAM1
- H4F5|SERF1|FAM2A|4F5|SMAM1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 8293
- 8293
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together