Order Entry
Belgium
ContactUsLinkComponent
Anti-RUNX2 Rabbit Polyclonal Antibody
Anti-RUNX2 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-RUNX2 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-77462SS
  • NOVUNBP1-77462
  • Antigen symbol:
  • RUNX2
  • RUNX2
  • Antigen name:
  • Runt-related Transcription Factor 2
  • Runt-related Transcription Factor 2
  • Antigen synonyms:
  • Acute myeloid leukemia 3 protein|PEBP2A|CBFA1|PEBP2-alpha A|SL3/AKV core-binding factor alpha A subunit|Polyomavirus enhancer-binding protein 2 alpha A subunit|OSF2|OSF-2|Core-binding factor subunit alpha-1|RUNX2|Runt-related transcription factor 2|PEA2-alpha A|CBF-alpha-1|Osteoblast-specific transcription factor 2|SL3-3 enhancer factor 1 alpha A subunit|AML3|Oncogene AML-3
  • Acute myeloid leukemia 3 protein|PEBP2A|CBFA1|PEBP2-alpha A|SL3/AKV core-binding factor alpha A subunit|Polyomavirus enhancer-binding protein 2 alpha A subunit|OSF2|OSF-2|Core-binding factor subunit alpha-1|RUNX2|Runt-related transcription factor 2|PEA2-alpha A|CBF-alpha-1|Osteoblast-specific transcription factor 2|SL3-3 enhancer factor 1 alpha A subunit|AML3|Oncogene AML-3
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 860
  • 860
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together