Order Entry
Belgium
ContactUsLinkComponent
 
Anti-LAMTOR2 Rabbit Polyclonal Antibody
  NOVUNBP1-71687
undefined
Anti-LAMTOR2 Rabbit Polyclonal Antibody
  NOVUNBP1-71687

 

  • Antibody type:
    Primary
  • Antigen name:
    late endosomal/lysosomal adaptor, MAPK and MTOR activator 2
  • Antigen symbol:
    ROBLD3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Gene ID:
    28956
  • Antigen synonyms:
    HSPC003|Ragulator2|ENDAP|p14|MAPBPIP|MAPKSP1AP|ROBLD3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µL
  • Pk:
    50 µl

 

 

The ROBLD3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ROBLD3. This antibody reacts with human, mouse. The ROBLD3 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:200

Type: Primary
Antigen: ROBLD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse