Order Entry
Belgium
ContactUsLinkComponent
 
Anti-SNX27 Rabbit Polyclonal Antibody
 
undefined
Anti-SNX27 Rabbit Polyclonal Antibody

 

  • Catalog No:
  • NOVUNBP1-45282
  • NOVUNBP1-45282SS
  • Antigen symbol:
  • SNX27
  • SNX27
  • Antigen name:
  • Sorting Nexin 27
  • Sorting Nexin 27
  • Antigen synonyms:
  • MRT1|MY014
  • MRT1|MY014
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 81609
  • 81609
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together