Order Entry
Belgium
ContactUsLinkComponent
 
Anti-Blimp-1 Mouse Monoclonal Antibody [clone: 3H2-E8]
 
Anti-Blimp-1 Mouse Monoclonal Antibody [clone: 3H2-E8]

 

  • Catalog No:
  • NOVUNB600-235SS
  • NOVUNB600-235
  • Antigen symbol:
  • BLIMP1
  • BLIMP1
  • Antigen name:
  • PR Domain Zinc Finger Protein 1
  • PR Domain Zinc Finger Protein 1
  • Antigen synonyms:
  • PRDI-BF1|B-lymphocyte-induced maturation protein 1|PR domain zinc finger protein 1|RP1-134E15.1|BLIMP1|BLIMP-1|PRDI-binding factor-1|beta-interferon gene positive-regulatory domain I binding factor
  • PRDI-BF1|B-lymphocyte-induced maturation protein 1|PR domain zinc finger protein 1|RP1-134E15.1|BLIMP1|BLIMP-1|PRDI-binding factor-1|beta-interferon gene positive-regulatory domain I binding factor
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 3H2-E8
  • 3H2-E8
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 639
  • 639
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together