Order Entry
Belgium
ContactUsLinkComponent
 
Anti-BIRC5 Mouse Monoclonal Antibody [clone: 32.1]
 
undefined
Anti-BIRC5 Mouse Monoclonal Antibody [clone: 32.1]

 

  • Catalog No:
  • NOVUNB500-237SS
  • NOVUNB500-237
  • Antigen symbol:
  • BIRC5
  • BIRC5
  • Antigen name:
  • Baculoviral IAP Repeat Containing 5
  • Baculoviral IAP Repeat Containing 5
  • Antigen synonyms:
  • Apoptosis Inhibitor 4|SVV|TIAP|IAP4|API4|Baculoviral IAP repeat containing 5|EPR 1|Apoptosis inhibitor survivin|BIRC 5
  • Apoptosis Inhibitor 4|SVV|TIAP|IAP4|API4|Baculoviral IAP repeat containing 5|EPR 1|Apoptosis inhibitor survivin|BIRC 5
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 32.1
  • 32.1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 332
  • 332
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together