Order Entry
Belgium
ContactUsLinkComponent
Anti-CHAF1B Mouse Monoclonal Antibody [clone: SS 53]
Anti-CHAF1B Mouse Monoclonal Antibody [clone: SS 53]
Supplier:  Novus Biologicals
Certificates
Anti-CHAF1B Mouse Monoclonal Antibody [clone: SS 53]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB500-212
  • NOVUNB500-212SS
  • Antigen symbol:
  • CHAF1B
  • CHAF1B
  • Antigen name:
  • Chromatin Assembly Factor 1, Subunit B (p60)
  • Chromatin Assembly Factor 1, Subunit B (p60)
  • Antigen synonyms:
  • CAF-1|MPHOSPH7|CAF1P60|CAF-IP60|MPP7|CAF1A|CAF1
  • CAF-1|MPHOSPH7|CAF1P60|CAF-IP60|MPP7|CAF1A|CAF1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • SS 53
  • SS 53
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 8208
  • 8208
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together