Anti-ROBO4 Rabbit Polyclonal Antibody
The ROBO4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ROBO4. This antibody reacts with human. The ROBO4 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml
Type: Primary
Antigen: ROBO4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- NOVUNB110-58780SS
- NOVUNB110-58780
- Antigen symbol:
- ROBO4
- ROBO4
- Antigen name:
- Roundabout Homolog 4
- Roundabout Homolog 4
- Antigen synonyms:
- MRB|MGC133352|MGC133353|UNQ421/PRO3674|FLJ20798|ECSM4
- MRB|MGC133352|MGC133353|UNQ421/PRO3674|FLJ20798|ECSM4
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 54538
- 54538
- Western blot:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together