Order Entry
Belgium
ContactUsLinkComponent
 
Anti-BMAL1 Rabbit Polyclonal Antibody
 
undefined
Anti-BMAL1 Rabbit Polyclonal Antibody

 

  • Catalog No:
  • NOVUNB100-2288
  • NOVUNB100-2288SS
  • Antigen symbol:
  • BMAL1
  • BMAL1
  • Antigen name:
  • Ayl hydrocarbon Receptor nuclear translocator-like
  • Ayl hydrocarbon Receptor nuclear translocator-like
  • Antigen synonyms:
  • ARNTL, BMAL1c, JAP3, MGC47515, MOP3, PASD3, TIC, ARNT-like protein 1, brain and muscle, bHLH-PAS protein JAP3, basic-helix-loop-helix-PAS orphan MOP3, member of PAS superfamily 3
  • ARNTL, BMAL1c, JAP3, MGC47515, MOP3, PASD3, TIC, ARNT-like protein 1, brain and muscle, bHLH-PAS protein JAP3, basic-helix-loop-helix-PAS orphan MOP3, member of PAS superfamily 3
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 406
  • 406
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • ChIP:
  • Yes
  • Yes
  • Storage buffer:
  • PBS
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together