Anti-APEX1 Mouse Monoclonal Antibody [clone: 13B8E5C2]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: APEX1 (APEX nuclease (multifunctional DNA repair enzyme) 1)
Clonality: monoclonal
Clone: 13B8E5C2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Specifications
- Catalog No:
- NOVUNB100-116
- NOVUNB100-116SS
- Antigen symbol:
- APEX1
- APEX1
- Antigen name:
- APEX nuclease (multifunctional DNA repair enzyme) 1
- APEX nuclease (multifunctional DNA repair enzyme) 1
- Antigen synonyms:
- Protein REF-1|APEX|DNA-(apurinic or apyrimidinic site) lyase|APEN|REF1|APE|APEX nuclease|APX|AP endonuclease 1|Apurinic-apyrimidinic endonuclease 1|APEX1|HAP1
- Protein REF-1|APEX|DNA-(apurinic or apyrimidinic site) lyase|APEN|REF1|APE|APEX nuclease|APX|AP endonuclease 1|Apurinic-apyrimidinic endonuclease 1|APEX1|HAP1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 13B8E5C2
- 13B8E5C2
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 328
- 328
- Isotype:
- IgG2b
- IgG2b
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Gel shift:
- Yes
- Yes
- ChIP:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together