Order Entry
Belgium
ContactUsLinkComponent
 
Anti-LCN1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1659
Supplier:  Boster Bio
undefined
Anti-LCN1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1659
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Lipocalin 1
  • Antigen symbol:
    LCN1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    Tlc|Lipocalin-1|TP VEG protein|VEGP|Lipocalin 1 like 2|LCN1|PMFA|Tear lipocalin|Tear prealbumin|lipocalin 1|Lipocalin 1 protein migrating faster than albumin|Von Ebner gland protein.|tear prealbumin|LCN1_HUMAN
  • Amino acid number:
    110 - 124
  • Sequence:
    HVKDHYIFYCEGELH
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of human LCN1 (110-124aa HVKDHYIFYCEGELH).
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Lipocalin-1(LCN1) detection. Tested with WB, IHC-P in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: LCN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human