Anti-RUNX2 Rabbit Polyclonal Antibody
Catalog # BSBTPA1224
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Runt-related Transcription Factor 2
- Antigen symbol:RUNX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:Acute myeloid leukemia 3 protein|PEBP2A|CBFA1|PEBP2-alpha A|SL3/AKV core-binding factor alpha A subunit|Polyomavirus enhancer-binding protein 2 alpha A subunit|OSF2|OSF-2|Core-binding factor subunit alpha-1|RUNX2|Runt-related transcription factor 2|PEA2-alpha A|CBF-alpha-1|Osteoblast-specific transcription factor 2|SL3-3 enhancer factor 1 alpha A subunit|AML3|Oncogene AML-3
- Amino acid number:244 - 258
- Sequence:DRLSDLGRIPHPSMR
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human RUNX2 (244-258aa DRLSDLGRIPHPSMR), identical to the related rat and mouse sequences
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Runt-related transcription factor 2 (RUNX2) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: RUNX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat