Order Entry
Belgium
ContactUsLinkComponent
Anti-NMDAR2B Rabbit Polyclonal Antibody
Anti-NMDAR2B Rabbit Polyclonal Antibody
Catalog # BSBTPA1059
Supplier:  Boster Bio
undefined
Anti-NMDAR2B Rabbit Polyclonal Antibody
Catalog # BSBTPA1059
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glutamate Receptor Ionotropic N-methyl D-aspartate 2B
  • Antigen symbol:
    NMDAR2B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Antigen synonyms:
    hNR3|MGC142178|GRIN2B|NR3|MGC142180|NR2B|Glutamate Receptor ionotropic N-methyl D-aspartate 2B|NMDAR2B
  • Amino acid number:
    1131 - 1146
  • Sequence:
    DFYLDQFRTKENSPHW
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human NMDAR2B (1131-1146aa, DFYLDQFRTKENSPHW), identical to the related mouse and rat sequence
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Glutamate receptor ionotropic, NMDA 2B (GRIN2B) detection. Tested with WB in Human;Mouse;Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: NMDAR2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat