Order Entry
Belgium
ContactUsLinkComponent
Anti-NMDAR1 Rabbit Monoclonal Antibody [clone: EPR2480Y]
Anti-NMDAR1 Rabbit Monoclonal Antibody [clone: EPR2480Y]
Supplier:  Abcam
New Product
Certificates
Anti-NMDAR1 Rabbit Monoclonal Antibody [clone: EPR2480Y]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCAAB68144-1ML
  • ABCAAB68144-100
  • ABCAAB68144-20UL
  • Antigen symbol:
  • GRIN1
  • Antigen name:
  • Glutamate receptor, ionotropic, N-methyl D-aspartate 1
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • EPR2480Y
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • Western blot:
  • Yes
  • ImmunoChemistry:
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

Specifications

Frequently Bought Together