Order Entry
Belgium
ContactUsLinkComponent
 
Anti-MDM2 Mouse Monoclonal Antibody [clone: 2A10]
Catalog # ABCAAB16895-50
Supplier:  Abcam
undefined
Anti-MDM2 Mouse Monoclonal Antibody [clone: 2A10]
Catalog # ABCAAB16895-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MDM2 oncogene, E3 ubiquitin Protein ligase
  • Antigen symbol:
    MDM2
  • Clonality:
    Monoclonal
  • Clone:
    2A10
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Seq:
    1202
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pack type:
    Vial
  • Pk:
    50 µG

Specifications

About this item

Mouse monoclonal [2A10] to MDM2.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: MDM2
Clonality: Monoclonal
Clone: 2A10
Conjugation:
Epitope: Within amino acids 294 - 339
Host: Mouse
Isotype: IgG2a
Reactivity: Human