Order Entry
Belgium
ContactUsLinkComponent
Anti-pan Cytokeratin Mouse Monoclonal Antibody [clone: C-11]
Anti-pan Cytokeratin Mouse Monoclonal Antibody [clone: C-11]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-pan Cytokeratin Mouse Monoclonal Antibody [clone: C-11]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCAAB7753-100
  • ABCAAB7753-1MG
  • Antigen name:
  • pan Cytokeratin
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • C-11
  • Host:
  • Mouse
  • Western blot:
  • Yes
  • ImmunoChemistry:
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Flow cytometry:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

Specifications

Frequently Bought Together