Anti-Hepatitis C Virus Core Antigen Mouse Monoclonal Antibody [clone: 6A1]
Catalogus # ABCAAB2582-100
Leverancier: Abcam
Specificaties
- Antibody type:Primary
- Antigen name:Hepatitis C Virus Core Antigen
- Clonality:Monoclonal
- Clone:1E5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Seq:1286
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pack type:Vial
- Pk:100 µG
Specificaties
Over dit artikel
Mouse monoclonal [6A1] to Hepatitis C Virus Core Antigen.
Let op: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Hepatitis C Virus Core Antigen
Clonality: Monoclonal
Clone: 6A1
Conjugation:
Epitope: This antibody recognises amino acid residues 1-80 of Hepatitis C Core Antigen
Host: Mouse
Isotype: IgG2a
Reactivity: Hepatitis C virus (HCV)